4-Nitro-o-phenylenediamine, 98%, Thermo Scientific Chemicals
4-Nitro-o-phenylenediamine, 98%, Thermo Scientific Chemicals
4-Nitro-o-phenylenediamine, 98%, Thermo Scientific Chemicals
4-Nitro-o-phenylenediamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitro-o-phenylenediamine, 98%, Thermo Scientific Chemicals
CAS: 99-56-9 | C6H7N3O2 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR148841000
Price (MYR)
1,011.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,011.00
Add to cart
Chemical Identifiers
CAS5351-23-5
IUPAC Name4-hydroxybenzohydrazide
Molecular FormulaC7H8N2O2
InChI KeyZMZGIVVRBMFZSG-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH>=97.5 %
Appearance (Color)White to light beige