Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ACR148805000
Price (MYR)
2,431.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
2,431.00
Add to cart
Chemical Identifiers
CAS1305-78-8
IUPAC Nameoxocalcium
Molecular FormulaCaO
InChI KeyODINCKMPIJJUCX-UHFFFAOYSA-N
SMILESO=[Ca]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentother alkaline earth and alkali metals 130ppm max
Total Metal Impurities0.008% max.