Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number ACR148801000
Price (MYR)
792.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
792.00
Add to cart
Chemical Identifiers
CAS26107-82-4
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)128.0-138.0?C
Appearance (Color)White to pale brown
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%