Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allylthiourea, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number ACR148800250
Price (MYR)
436.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
436.00
Add to cart
Chemical Identifiers
CAS109-57-9
IUPAC Name(prop-2-en-1-yl)thiourea
Molecular FormulaC4H8N2S
InChI KeyHTKFORQRBXIQHD-UHFFFAOYSA-N
SMILESNC(=S)NCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Melting point70°C to 78°C
Loss on drying=<1.5 % (50°C, 2 hrs)
Infrared spectrumConforms
Appearance (Form)Powder or crystals or granules and/or chunks