2-Cyanoacetamide, 99%, Thermo Scientific Chemicals
2-Cyanoacetamide, 99%, Thermo Scientific Chemicals
2-Cyanoacetamide, 99%, Thermo Scientific Chemicals
2-Cyanoacetamide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Cyanoacetamide, 99%, Thermo Scientific Chemicals
CAS: 107-91-5 | C3H4N2O | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number ACR148655000
Price (MYR)
919.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
919.00
Add to cart
Specifications
Chemical Name or Material2-Cyanoacetamide
Name Note99%
CAS107-91-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning