Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ACR148535000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetaldehyde ammonia trimer
Name Note98%
CAS58052-80-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning