Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number ACR148530250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2708-97-6
IUPAC Name1,2,3,4-tetrafluoro-5,6-diiodobenzene
Molecular FormulaC6F4I2
InChI KeyJQBYIZAYQMMVTO-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(I)C(I)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Melting point49°C to 53°C
Appearance (Color)White to off-white
GC>=98.5 %