Nitromethane, 99%, pure, Thermo Scientific Chemicals
Nitromethane, 99%, pure, Thermo Scientific Chemicals
Nitromethane, 99%, pure, Thermo Scientific Chemicals
Nitromethane, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitromethane, 99%, pure, Thermo Scientific Chemicals
CAS: 75-52-5 | CH3NO2 | 61.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR148510010
Price (MYR)
1,052.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,052.00
Add to cart
Chemical Identifiers
CAS58-61-7
IUPAC Name(2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular FormulaC10H13N5O4
InChI KeyOIRDTQYFTABQOQ-KQYNXXCUSA-N
SMILESNC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Optical Rotation-70.0 ± 2.0? (C=2 in 5% NaOH)