Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
CAS: 98-10-2 | C6H7NO2S | 157.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number ACR148495000
Price (MYR)
706.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
706.00
Add to cart
Chemical Identifiers
CAS92112-69-1
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at300nm: ≤0.005AU
Residue on Evaporation≤5 mg/ml
Assay (GC)≥60.0% as n-Hexane
Assay (GC)≥99%, as C6 Hydrocarbons
Optical Absorbance at195nm: ≤1.00AU