Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
CAS: 98-10-2 | C6H7NO2S | 157.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number ACR148491000
Price (MYR)
401.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
401.00
Add to cart
Chemical Identifiers
CAS374538-04-2
IUPAC Name(4-cyclohexylphenyl)boronic acid
Molecular FormulaC12H17BO2
InChI KeyKNQVRFYNQWNYPU-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
Assay (Aqueous acid-base Titration)≥97.5%
FormCrystals or powder or crystalline powder
Proton NMRConforms to structure
Assay (HPLC)≥97.5%