Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzenesulfonamide, 98+%, Thermo Scientific Chemicals
CAS: 98-10-2 | C6H7NO2S | 157.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number ACR148490050
Price (MYR)
378.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
378.00
Add to cart
Specifications
Chemical Name or MaterialBenzenesulfonamide
Name Note98+%
CAS98-10-2
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning