Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number ACR148482500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS537-45-1
IUPAC Name2,6-dibromo-4-(chloroimino)cyclohexa-2,5-dien-1-one
Molecular FormulaC6H2Br2ClNO
InChI KeyJYWKEVKEKOTYEX-UHFFFAOYSA-N
SMILESClN=C1C=C(Br)C(=O)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline needles
Appearance (Color)Yellow to ochre
AppearanceMay darken on storage
Infrared spectrumConforms
Melting point76°C to 83°C