Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number ACR148480010
Price (MYR)
1,065.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,065.00
Add to cart
Chemical Identifiers
CAS69-65-8
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<0.3 % (105°C, 4 hrs)
Specific optical rotation+23° to +25° (20°C, 589 nm) (Ph.Eur.-directives)
HPLC>=98.0 %
Heavy metals (as Pb)=<5 ppm