Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nicotinic acid hydrazide, 97%, Thermo Scientific Chemicals
CAS: 553-53-7 | C6H7N3O | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number ACR148200250
Price (MYR)
543.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
543.00
Add to cart
Specifications
Chemical Name or MaterialNicotinic acid hydrazide
Name Note97%
CAS553-53-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning