2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
CAS: 4767-03-7 | C5H10O4 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR147975000
Price (MYR)
578.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
578.00
Add to cart
Specifications
Chemical Name or Material2, 2-Bis(hydroxymethyl)propionic acid
CAS4767-03-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,