2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)propionic acid, 98%, Thermo Scientific Chemicals
CAS: 4767-03-7 | C5H10O4 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
Plastic bottle
Catalog number ACR147970010
Price (MYR)
908.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
908.00
Add to cart
Chemical Identifiers
CAS25322-69-4
Molecular Formula(C3H6O)n
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES[H]OC(C)CO
MDL NumberMFCD00084446
SpecificationsSpecification SheetSpecification Sheet
Hydroxyl Value50 - 60mg KOH/g
Refractive Index1.4490 - 1.4530 @ 20?C
FormLiquid
Appearance (Color)Clear, colorless
Water Content (Karl Fischer Titration)< 0.10%