Formic acid, 98+%, pure, Thermo Scientific Chemicals
Formic acid, 98+%, pure, Thermo Scientific Chemicals
Formic acid, 98+%, pure, Thermo Scientific Chemicals
Formic acid, 98+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formic acid, 98+%, pure, Thermo Scientific Chemicals
CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Plastic bottle
Catalog number ACR147930010
Price (MYR)
926.00
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
926.00
Add to cart
Chemical Identifiers
CAS18997-19-8
IUPAC Namechloromethyl 2,2-dimethylpropanoate
Molecular FormulaC6H11ClO2
InChI KeyGGRHYQCXXYLUTL-UHFFFAOYSA-N
SMILESCC(C)(C)C(=O)OCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4160 to 1.4180 (20°C, 589 nm)
GC>=99.0 %