1-Dodecanesulfonic acid, sodium salt, 99+%
1-Dodecanesulfonic acid, sodium salt, 99+%
1-Dodecanesulfonic acid, sodium salt, 99+%
1-Dodecanesulfonic acid, sodium salt, 99+%
Thermo Scientific Chemicals
1-Dodecanesulfonic acid, sodium salt, 99+%
CAS: 2386-53-0 | C12H25NaO3S | 272.38 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1479000101 gGlass bottle
Catalog number ACR147900010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS100-44-7
IUPAC Name(chloromethyl)benzene
Molecular FormulaC7H7Cl
InChI KeyKCXMKQUNVWSEMD-UHFFFAOYSA-N
SMILESClCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Stabilizer0.25 % propylene oxide
Infrared spectrumConforms
Refractive index1.5370 to 1.5390 (20°C, 589 nm)
Appearance (Form)Liquid