Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, trisodium salt hydrate, 98%
CAS: 85715-60-2 | C10H13N2Na3O8 | 358.19 g/mol
Have Questions?
Catalog number ACR147865000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylenediaminetetraacetic acid, trisodium salt hydrate
Name Note98%
CAS85715-60-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
View more
Chelation/Complexation reagent
RUO – Research Use Only