Salicylic Acid, 99+%, Thermo Scientific Chemicals
Salicylic Acid, 99+%, Thermo Scientific Chemicals
Salicylic Acid, 99+%, Thermo Scientific Chemicals
Salicylic Acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Salicylic Acid, 99+%, Thermo Scientific Chemicals
CAS: 69-72-7 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number ACR147700010
Price (MYR)
714.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
714.00
Add to cart
Chemical Identifiers
CAS1483-72-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder