Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Plastic Drum
Catalog number ACR146750250
Price (MYR)
5,811.00
Add to cart
Quantity:
25 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
5,811.00
Add to cart
Chemical Identifiers
CAS1309-48-4
IUPAC Nameoxomagnesium
Molecular FormulaMgO
InChI KeyCPLXHLVBOLITMK-UHFFFAOYSA-N
SMILESO=[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.