Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number ACR146750025
Price (MYR)
846.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
846.00
Add to cart
Chemical Identifiers
CAS87-89-8
IUPAC Namecyclohexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H12O6
InChI KeyCDAISMWEOUEBRE-UHFFFAOYSA-N
SMILESOC1C(O)C(O)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Loss on Drying≤ 0.5 %
Appearance (Color)White
Assay (HPLC)≥ 97.0 %
Heavy Metals≤ 10 ppm