1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
CAS: 119-64-2 | C10H12 | 132.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number ACR146730025
Price (MYR)
1,380.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,380.00
Add to cart
Chemical Identifiers
CAS18113-03-6
IUPAC Name2-chloro-4-methoxyphenol
Molecular FormulaC7H7ClO2
InChI KeyGNVRRKLFFYSLGT-UHFFFAOYSA-N
SMILESCOC1=CC=C(O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)37-44?C
FormCrystalline powder or Fused solid
Appearance (Color)White to cream to yellow to brown
Assay (GC)≥96.0%