1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,3,4-Tetrahydronaphthalene, 98+%, pure, Thermo Scientific Chemicals
CAS: 119-64-2 | C10H12 | 132.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number ACR146730010
Price (MYR)
873.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
873.00
Add to cart
Chemical Identifiers
CAS1694-92-4
IUPAC Name2-nitrobenzene-1-sulfonyl chloride
Molecular FormulaC6H4ClNO4S
InChI KeyWPHUUIODWRNJLO-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to brown
Infrared spectrumConforms
GC>=96.0 %
Melting point64°C to 70°C
Appearance (Form)Crystalline powder