Cyclohexanemethylamine, 97%, Thermo Scientific Chemicals
Cyclohexanemethylamine, 97%, Thermo Scientific Chemicals
Cyclohexanemethylamine, 97%, Thermo Scientific Chemicals
Cyclohexanemethylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanemethylamine, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR146570250
Price (MYR)
2,291.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,291.00
Add to cart
Specifications
Chemical Name or MaterialCyclohexanemethylamine
CAS3218-02-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air