1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals
CAS: 637-87-6 | C6H4ClI | 238.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR146480250
Price (MYR)
542.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
542.00
Add to cart
Specifications
Chemical Name or Material1-Chloro-4-iodobenzene
Name Note99%
CAS637-87-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning