Triphenylmethane, 99+%, Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
CAS: 519-73-3 | C19H16 | 244.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR146385000
Price (MYR)
2,536.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,536.00
Add to cart
Chemical Identifiers
CAS638-67-5
IUPAC Nametricosane
Molecular FormulaC23H48
InChI KeyFIGVVZUWCLSUEI-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point47°C to 50°C
Appearance (Form)Crystals, flakes and/or chunks
GC>=97.5 %
Appearance (Color)White