Triphenylmethane, 99+%, Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenylmethane, 99+%, Thermo Scientific Chemicals
CAS: 519-73-3 | C19H16 | 244.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR146380250
Price (MYR)
488.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
488.00
Add to cart
Chemical Identifiers
CAS652148-91-9
IUPAC Name(5-chloropyridin-2-yl)boronic acid
Molecular FormulaC5H5BClNO2
InChI KeyJEGHCYRKSUGHJH-UHFFFAOYSA-N
SMILESOB(O)C1=NC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%