2-Bromo-4'-phenylacetophenone, 98%, Thermo Scientific Chemicals
2-Bromo-4'-phenylacetophenone, 98%, Thermo Scientific Chemicals
2-Bromo-4'-phenylacetophenone, 98%, Thermo Scientific Chemicals
2-Bromo-4'-phenylacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-4'-phenylacetophenone, 98%, Thermo Scientific Chemicals
CAS: 135-73-9 | C14H11BrO | 275.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR146350250
Price (MYR)
1,176.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,176.00
Add to cart
Chemical Identifiers
CAS12033-89-5
IUPAC NameSilicon nitride
Molecular FormulaN4Si3
InChI KeyHQVNEWCFYHHQES-UHFFFAOYSA-N
SMILESN12[Si]34N5[Si]11N3[Si]25N41
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey or brown
FormPowder
Assay from Suppliers CofAAlpha phase: ≥87.0%
Particle Size≤10µm