1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
Thermo Scientific Chemicals
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Packaging:
Glass bottle
Catalog number ACR146310500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS80-70-6
IUPAC NameN,N,N',N'-tetramethylguanidine
Molecular FormulaC5H13N3
InChI KeyKYVBNYUBXIEUFW-UHFFFAOYSA-N
SMILESCN(C)C(=N)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Refractive index1.4665 to 1.4695 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Indicator
RUO – Research Use Only