Para Red, pure
Para Red, pure
Para Red, pure
Thermo Scientific Chemicals
Para Red, pure
C16H11N3O3
Have Questions?
Catalog NumberQuantity
ACR14630025025 g
Catalog number ACR146300250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS22572-40-3
IUPAC Name(3-{[(ethylimino)methylidene]amino}propyl)trimethylazanium iodide
Molecular FormulaC9H20IN3
InChI KeyAGSKWMRPXWHSPF-UHFFFAOYSA-M
SMILES[I-].CCN=C=NCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Iodide)≥97.5 to ≤102.5%
Melting Point (clear melt)91-97?C
FormCrystalline powder
Appearance (Color)White to cream to yellow
Water Content (Karl Fischer Titration)≤1.0%