Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urea, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
1 kg
5 kg
Packaging:
View all
Plastic bag in resealable cardbox
Catalog number ACR140750250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 kg
Packaging:
Plastic bag in resealable cardbox
Request bulk or custom format
Specifications
Chemical Name or MaterialUrea
CAS57-13-6
Melting Point131.0°C to 135.0°C
ColorWhite
EINECS Number200-315-5
View more
For analysis
RUO – Research Use Only