DL-Tropic acid, 97%, Thermo Scientific Chemicals
DL-Tropic acid, 97%, Thermo Scientific Chemicals
DL-Tropic acid, 97%, Thermo Scientific Chemicals
DL-Tropic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Tropic acid, 97%, Thermo Scientific Chemicals
CAS: 552-63-6 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR140551000
Price (MYR)
2,125.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,125.00
Add to cart
Specifications
Chemical Name or MaterialDL-Tropic acid
CAS552-63-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point117.0°C to 120.0°C