Trithiocyanuric acid, 95%, Thermo Scientific Chemicals
Trithiocyanuric acid, 95%, Thermo Scientific Chemicals
Trithiocyanuric acid, 95%, Thermo Scientific Chemicals
Trithiocyanuric acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trithiocyanuric acid, 95%, Thermo Scientific Chemicals
CAS: 638-16-4 | C3H3N3S3 | 177.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR140530050
Price (MYR)
453.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
453.00
Add to cart
Chemical Identifiers
CAS75-65-0
IUPAC Name2-methylpropan-2-ol
Molecular FormulaC4H10O
InChI KeyDKGAVHZHDRPRBM-UHFFFAOYSA-N
SMILESCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.3860 to 1.3880 (20°C, 589 nm)
Infrared spectrumConforms
Water=<0.03 % (K.F.)
Appearance (Form)Liquid after melting