Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
CAS: 101-02-0 | C18H15O3P | 310.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass Bottle
Catalog number ACR140440025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS141-82-2
IUPAC Namepropanedioic acid
Molecular FormulaC3H4O4
InChI KeyOFOBLEOULBTSOW-UHFFFAOYSA-N
SMILESOC(=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with NaOH>=98.5 %
Melting point133°C to 136°C
Sulfate (SO4)=<500 ppm
Chloride (Cl)=<500 ppm