Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenyl phosphite, 99%, Thermo Scientific Chemicals
CAS: 101-02-0 | C18H15O3P | 310.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number ACR140440010
Price (MYR)
689.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
689.00
Add to cart
Specifications
Chemical Name or MaterialTriphenyl phosphite
Acidity1.5mg KOH/g max.
CAS101-02-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.