Triphenylphosphine oxide, 99%, Thermo Scientific Chemicals
Triphenylphosphine oxide, 99%, Thermo Scientific Chemicals
Triphenylphosphine oxide, 99%, Thermo Scientific Chemicals
Triphenylphosphine oxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenylphosphine oxide, 99%, Thermo Scientific Chemicals
CAS: 791-28-6 | C18H15OP | 278.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR140431000
Price (MYR)
625.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
625.00
Add to cart
Chemical Identifiers
CAS502496-27-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown
FormCrystals or powder or crystalline powder or flakes
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
Melting Point (clear melt)260.5-269.5?C