Triphenylmethyl chloride, 98%, Thermo Scientific Chemicals
Triphenylmethyl chloride, 98%, Thermo Scientific Chemicals
Triphenylmethyl chloride, 98%, Thermo Scientific Chemicals
Triphenylmethyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenylmethyl chloride, 98%, Thermo Scientific Chemicals
CAS: 76-83-5 | C19H15Cl | 278.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR140401000
Price (MYR)
449.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
449.00
Add to cart
Specifications
Chemical Name or MaterialTriphenylmethyl chloride
Name Note98%
CAS76-83-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.