Triphenylene, 98%, Thermo Scientific Chemicals
Triphenylene, 98%, Thermo Scientific Chemicals
Triphenylene, 98%, Thermo Scientific Chemicals
Triphenylene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triphenylene, 98%, Thermo Scientific Chemicals
CAS: 217-59-4 | C18H12 | 228.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR140360010
Price (MYR)
611.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
611.00
Add to cart
Specifications
Chemical Name or MaterialTriphenylene
Name Note98%
CAS217-59-4
Health Hazard 1GHS Signal Word: Danger
Melting Point196.0°C to 200.0°C