2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 527-60-6 | C9H12O | 136.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number ACR140210500
Price (MYR)
1,059.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,059.00
Add to cart
Chemical Identifiers
CAS9048-46-8
MDL NumberMFCD00145743
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Residual water≤5.0%
Assay (unspecified)Fatty Acid: ≤0.1 mg/gm
Appearance (Color)Pale yellow to cream
Assay from Supplier's CofA≥97.5% (protein electrophoresis)