2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 527-60-6 | C9H12O | 136.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number ACR140210100
Price (MYR)
468.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
468.00
Add to cart
Chemical Identifiers
CAS1338-39-2
IUPAC Name2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl dodecanoate
Molecular FormulaC18H34O6
InChI KeyLWZFANDGMFTDAV-IOVMHBDKSA-N
SMILESCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSaponification number 157-171
pHpH <8.0
FormViscous liquid
Identification (FTIR)Conforms
Refractive Index1.4710-1.4770 @ 20?C