4',5,7-Trihydroxyflavanone, 97%
4',5,7-Trihydroxyflavanone, 97%
4',5,7-Trihydroxyflavanone, 97%
4',5,7-Trihydroxyflavanone, 97%
Thermo Scientific Chemicals
4',5,7-Trihydroxyflavanone, 97%
CAS: 480-41-1 | C15H12O5 | 272.26 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR13982010010 gGlass bottle
Catalog number ACR139820100
Price (MYR)
1,056.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,056.00
Add to cart
Chemical Identifiers
CAS31551-45-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥96.0% (based on elemental N)
Assay from Supplier's CofA≥96.0% (HPLC)