Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Catalog number ACR139720250
Price (MYR)
504.00
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
504.00
Add to cart
Chemical Identifiers
CAS67-48-1
IUPAC Name(2-hydroxyethyl)trimethylazanium chloride
Molecular FormulaC5H14ClNO
InChI KeySGMZJAMFUVOLNK-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or needles or adhering crystals
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤2%