Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
CAS: 867-13-0 | C8H17O5P | 224.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR139705000
Price (MYR)
1,952.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,952.00
Add to cart
Chemical Identifiers
CAS621-42-1
IUPAC NameN-(3-hydroxyphenyl)acetamide
Molecular FormulaC8H9NO2
InChI KeyQLNWXBAGRTUKKI-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pink to grey or pale yellow to pale orange to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)144.0-151.0°C
for C-C Bond Formation (Olefination)
RUO – Research Use Only