Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals
CAS: 867-13-0 | C8H17O5P | 224.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR139701000
Price (MYR)
695.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
695.00
Add to cart
Specifications
Chemical Name or MaterialTriethyl phosphonoacetate
Acidity2mg KOH/g max.
CAS867-13-0
Health Hazard 1GHS Signal Word: Danger
Melting Point-24.0°C
View more
for C-C Bond Formation (Olefination)
RUO – Research Use Only