Tricarballylic acid, 99%, Thermo Scientific Chemicals
Tricarballylic acid, 99%, Thermo Scientific Chemicals
Tricarballylic acid, 99%, Thermo Scientific Chemicals
Tricarballylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tricarballylic acid, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ACR1393600505 g
Catalog number ACR139360050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS30361-29-6
IUPAC Name(2E,4E)-undeca-2,4-dienal
Molecular FormulaC11H18O
InChI KeyUVIUIIFPIWRILL-XBLVEGMJSA-N
SMILESCCCCCCC=CC=CC=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5080-1.5160 @ 20?C
Assay (GC)>90.0% (major isomer)
Appearance (Color)Clear colorless to yellow