1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Plastic bottle
Catalog number ACR139280250
Price (MYR)
440.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
440.00
Add to cart
Chemical Identifiers
CAS100-22-1
IUPAC NameN1,N1,N4,N4-tetramethylbenzene-1,4-diamine
Molecular FormulaC10H16N2
InChI KeyCJAOGUFAAWZWNI-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)47.0-54.0?C
Appearance (Color)White to cream to red to dark brown to brown or pale blue to blue to purple to pale gray to gray
FormCrystals or powder or crystalline powder or lumps or fused solid or flakes or granules