2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number ACR139251000
Price (MYR)
1,516.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,516.00
Add to cart
Chemical Identifiers
CAS2181-42-2
IUPAC Nametrimethylsulfanium iodide
Molecular FormulaC3H9IS
InChI KeyVFJYIHQDILEQNR-UHFFFAOYSA-M
SMILES[I-].C[S+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (Titration ex Iodide)≥97.5 to ≤102.5%