2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR139250250
Price (MYR)
599.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
599.00
Add to cart
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofATitrable acid: ≤5 mL (≤0.001 meq/g, ≤0.056 mgKOH/g)
Water Content (Karl Fischer Titration)≤0.1%
FormLiquid
Assay from Supplier's CofA≥99.9% (purity by GC)
Appearance (Color)Clear colorless