Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
CAS: 102-76-1 | C9H14O6 | 218.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR139225000
Price (MYR)
582.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
582.00
Add to cart
Chemical Identifiers
CAS207801-28-3
IUPAC Nameyttrium(3+) triacetate
Molecular FormulaC6H9O6Y
InChI KeyJUWHRRAPBUAYTA-UHFFFAOYSA-K
SMILES[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Total conc. of imp.as REO=<0.1 %
Appearance (Color)White
Appearance (Form)Crystalline powder and/or lumps